Product Details
- SNP ID
-
rs150861980
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
2
- Location
-
Chr.1:156669807 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGGGGACACTGACACTCACAGAAT[C/T]AGACTCCAGGAATTCCCCTCTCTGG
- Phenotype
-
MIM: 600915
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NES
PubMed Links
Gene Details
- Gene
- NES
- Gene Name
- nestin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006617.1 |
4514 |
Missense Mutation |
AAT,GAT |
N1461D |
NP_006608.1 |
View Full Product Details