Product Details

SNP ID
rs151074666
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:97079056 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCTGGAAACCATTTTGATGCAGT[C/T]GACAATAGGGCAAACACTGAGACAC
Phenotype
MIM: 612779
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DPYD PubMed Links

Gene Details

Gene
DPYD
Gene Name
dihydropyrimidine dehydrogenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000110.3 3345 Missense Mutation AAC,GAC N1000D NP_000101.2
NM_001160301.1 3345 Intron NP_001153773.1
XM_005270562.3 3345 Missense Mutation AAC,GAC N928D XP_005270619.2
XM_006710397.3 3345 Intron XP_006710460.1
XM_017000507.1 3345 Missense Mutation AAC,GAC N963D XP_016855996.1
XM_017000508.1 3345 Missense Mutation AAC,GAC N835D XP_016855997.1
XM_017000509.1 3345 Missense Mutation AAC,GAC N835D XP_016855998.1
XM_017000510.1 3345 Missense Mutation AAC,GAC N835D XP_016855999.1

View Full Product Details