Product Details

SNP ID
rs138025844
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:59030421 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTTTTGCACAGATCTGGGAGTAT[C/G]GGATGTAGCTGGGAGAAAATGAGAG
Phenotype
MIM: 606153 MIM: 612901
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ATP5E PubMed Links

Gene Details

Gene
ATP5E
Gene Name
ATP synthase, H+ transporting, mitochondrial F1 complex, epsilon subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006886.3 Intron NP_008817.1
Gene
PRELID3B
Gene Name
PRELI domain containing 3B
There are no transcripts associated with this gene.

Gene
SLMO2-ATP5E
Gene Name
SLMO2-ATP5E readthrough
There are no transcripts associated with this gene.

Gene
TUBB1
Gene Name
tubulin beta 1 class VI
There are no transcripts associated with this gene.

View Full Product Details