Product Details
- SNP ID
-
rs140341486
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:45649257 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCAAGCAAACATCTCCCAACTCAC[A/G]ACGTTTATCCAGCAGATGTTTCCAC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
WFDC11
PubMed Links
Gene Details
- Gene
- WFDC11
- Gene Name
- WAP four-disulfide core domain 11
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_147197.2 |
438 |
Silent Mutation |
GTC,GTT |
V81V |
NP_671730.1 |
View Full Product Details