Product Details
- SNP ID
-
rs141436003
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:38750119 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGTGCTACGGGATTCCCAAGGTTG[C/T]CTATGGAATAGACAGCCTCTTCAGC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ACTR5
PubMed Links
Gene Details
- Gene
- ACTR5
- Gene Name
- ARP5 actin-related protein 5 homolog
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_024855.3 |
510 |
Missense Mutation |
GCC,GTC |
A162V |
NP_079131.3 |
View Full Product Details