Product Details

SNP ID
rs143943850
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:44197635 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGAGTTGACTGCTGGAAGAAGAC[A/G]CTATTGTACTGCAATGTATAAACTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OSER1 PubMed Links

Gene Details

Gene
OSER1
Gene Name
oxidative stress responsive serine rich 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016470.7 592 Missense Mutation GCG,GTG A99V NP_057554.4
XM_011528851.1 592 Missense Mutation GCG,GTG A99V XP_011527153.1
XM_011528853.2 592 Missense Mutation GCG,GTG A99V XP_011527155.1
XM_011528854.2 592 Missense Mutation GCG,GTG A61V XP_011527156.1
XM_017027873.1 592 Missense Mutation GCG,GTG A99V XP_016883362.1

View Full Product Details