Product Details

SNP ID
rs145864345
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:63256739 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTCCATCATTACACAGAATCCCA[C/T]GTGGGAAGACCCTAGTCGGAGGCCC
Phenotype
MIM: 612873
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FLJ16779 PubMed Links

Gene Details

Gene
FLJ16779
Gene Name
uncharacterized LOC100192386
There are no transcripts associated with this gene.

Gene
LOC107985425
Gene Name
uncharacterized LOC107985425
There are no transcripts associated with this gene.

Gene
NKAIN4
Gene Name
Na+/K+ transporting ATPase interacting 4
There are no transcripts associated with this gene.

View Full Product Details