Product Details

SNP ID
rs151193257
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:1980135 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTGTGGGGAAGGGACATCCACCCT[A/T]CCCCATCACAGACCCCAGAGAAATA
Phenotype
MIM: 131340
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC727993 PubMed Links

Gene Details

Gene
LOC727993
Gene Name
uncharacterized LOC727993
There are no transcripts associated with this gene.

Gene
PDYN
Gene Name
prodynorphin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190892.1 1228 UTR 3 NP_001177821.1
NM_001190898.2 1228 UTR 3 NP_001177827.1
NM_001190899.2 1228 UTR 3 NP_001177828.1
NM_001190900.1 1228 UTR 3 NP_001177829.1
NM_024411.4 1228 UTR 3 NP_077722.1
XM_011529244.1 1228 UTR 3 XP_011527546.1
XM_011529245.1 1228 UTR 3 XP_011527547.1
XM_011529246.2 1228 UTR 3 XP_011527548.1
XM_011529247.1 1228 UTR 3 XP_011527549.1
XM_011529248.1 1228 UTR 3 XP_011527550.1
XM_011529249.2 1228 UTR 3 XP_011527551.1
XM_011529250.2 1228 UTR 3 XP_011527552.1
XM_017027878.1 1228 UTR 3 XP_016883367.1

View Full Product Details