Product Details

SNP ID
rs148617415
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:29027952 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACAAAGGGAAAAACTGTTCCAAT[C/T]GATGATTCCTCTGAAACTTTAGGTA
Phenotype
MIM: 604735
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RWDD2B PubMed Links

Gene Details

Gene
RWDD2B
Gene Name
RWD domain containing 2B
There are no transcripts associated with this gene.

Gene
USP16
Gene Name
ubiquitin specific peptidase 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001992.1 242 Silent Mutation ATC,ATT I13I NP_001001992.1
NM_001032410.1 242 Silent Mutation ATC,ATT I13I NP_001027582.1
NM_006447.2 242 Silent Mutation ATC,ATT I13I NP_006438.1
XM_017028257.1 242 Silent Mutation ATC,ATT I13I XP_016883746.1
XM_017028258.1 242 Silent Mutation ATC,ATT I13I XP_016883747.1
XM_017028259.1 242 Silent Mutation ATC,ATT I13I XP_016883748.1
XM_017028260.1 242 Intron XP_016883749.1
XM_017028261.1 242 UTR 5 XP_016883750.1

View Full Product Details