Product Details

SNP ID
rs141336766
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:41913468 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTCCTCGTTCCAACCGAGGAAGG[A/G]GACCTGGACCCTGTAGTGCCCACTT
Phenotype
MIM: 600481
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR378I PubMed Links

Gene Details

Gene
MIR378I
Gene Name
microRNA 378i
There are no transcripts associated with this gene.

Gene
SHISA8
Gene Name
shisa family member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001207020.1 Intron NP_001193949.1
XM_006724254.3 Intron XP_006724317.1
XM_006724255.2 Intron XP_006724318.1
XM_006724256.3 Intron XP_006724319.1
Gene
SREBF2
Gene Name
sterol regulatory element binding transcription factor 2
There are no transcripts associated with this gene.

View Full Product Details