Product Details

SNP ID
rs141375308
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:44495483 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAACAGCACAGTCCAAAGGGCGTG[A/G]CCAAACTCAGCCACAAAATGATTTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LDOC1L PubMed Links

Gene Details

Gene
LDOC1L
Gene Name
leucine zipper down-regulated in cancer 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032287.2 2411 UTR 3 NP_115663.2

View Full Product Details