Product Details

SNP ID
rs146023535
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:21959382 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACGGCAGGGCAGCAGCCCTGTGC[A/G]TCAGAGGCACGTGTTCCTGGCCATG
Phenotype
MIM: 603582
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PPM1F PubMed Links

Gene Details

Gene
PPM1F
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1F
There are no transcripts associated with this gene.

Gene
TOP3B
Gene Name
topoisomerase (DNA) III beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282112.1 Intron NP_001269041.1
NM_001282113.1 Intron NP_001269042.1
NM_003935.4 Intron NP_003926.1
XM_005261811.1 Intron XP_005261868.1
XM_005261813.2 Intron XP_005261870.1
XM_006724349.1 Intron XP_006724412.1
XM_006724350.1 Intron XP_006724413.1
XM_011530482.1 Intron XP_011528784.1
XM_017029038.1 Intron XP_016884527.1
XM_017029039.1 Intron XP_016884528.1
XM_017029040.1 Intron XP_016884529.1
XM_017029041.1 Intron XP_016884530.1

View Full Product Details