Product Details

SNP ID
rs149218137
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:22245156 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCAGGGCCCCCAGGTCCCCCCTC[A/G]CTTCTCTGGATCCAAAGATGTGGCC
Phenotype
MIM: 605141
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
VPREB1 PubMed Links

Gene Details

Gene
VPREB1
Gene Name
pre-B lymphocyte 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303509.1 277 Missense Mutation CAC,CGC H85R NP_001290438.1
NM_007128.3 277 Missense Mutation CAC,CGC H86R NP_009059.1

View Full Product Details