Product Details

SNP ID
rs150118170
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:29058877 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGACTGCAAAGAGCCTCCCAGAG[C/T]TTTTGTTTAATGGCCTTGCCCAGCT
Phenotype
MIM: 612062
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C22orf31 PubMed Links

Gene Details

Gene
C22orf31
Gene Name
chromosome 22 open reading frame 31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015370.1 660 Silent Mutation AAA,AAG K246K NP_056185.1
XM_005261491.3 660 Silent Mutation AAA,AAG K142K XP_005261548.1
XM_011530096.2 660 Silent Mutation AAA,AAG K205K XP_011528398.1
XM_017028741.1 660 Silent Mutation AAA,AAG K205K XP_016884230.1
Gene
ZNRF3
Gene Name
zinc and ring finger 3
There are no transcripts associated with this gene.

View Full Product Details