Product Details
- SNP ID
-
rs138421941
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:21001973 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTAATCAGAAATTATTTTCTTCGT[C/T]GCAATGGCCTGGCTTTTAATTATTT
- Phenotype
-
MIM: 107730
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
APOB
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs12713450] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- APOB
- Gene Name
- apolipoprotein B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000384.2 |
13577 |
Silent Mutation |
GCA,GCG |
A4483A |
NP_000375.2 |
View Full Product Details