Product Details

SNP ID
rs138584387
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:178432160 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTGTGAGCTGCATCACTTTGTGC[A/G]TTGCCACAGGAGATACCGGAGCCAT
Phenotype
MIM: 603424
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101927027 PubMed Links

Gene Details

Gene
LOC101927027
Gene Name
uncharacterized LOC101927027
There are no transcripts associated with this gene.

Gene
PRKRA
Gene Name
protein activator of interferon induced protein kinase EIF2AK2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001139517.1 1121 Silent Mutation AAC,AAT N282N NP_001132989.1
NM_001139518.1 1121 Silent Mutation AAC,AAT N268N NP_001132990.1
NM_001316362.1 1121 Silent Mutation AAC,AAT N180N NP_001303291.1
NM_003690.4 1121 Silent Mutation AAC,AAT N293N NP_003681.1
XM_011512063.1 1121 Silent Mutation AAC,AAT N208N XP_011510365.1
XM_011512066.1 1121 Silent Mutation AAC,AAT N180N XP_011510368.1
XM_017005159.1 1121 Silent Mutation AAC,AAT N180N XP_016860648.1

View Full Product Details