Product Details

SNP ID
rs139094728
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:88028354 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGAATATTGCATGATCTTGGGTA[C/G]GTCTGGATGGTTTCAGATGGGAGTC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KRCC1 PubMed Links

Gene Details

Gene
KRCC1
Gene Name
lysine rich coiled-coil 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304526.1 977 Silent Mutation ACC,ACG T70T NP_001291455.1
NM_016618.2 977 Silent Mutation ACC,ACG T70T NP_057702.1
XM_017004292.1 977 Silent Mutation ACC,ACG T70T XP_016859781.1

View Full Product Details