Product Details

SNP ID
rs141844180
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:10441917 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAATACATAAAGGTCTGCTCACTC[A/G]ACTCATCTTCGTCTAGAAAGGCAGA
Phenotype
MIM: 165640
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101929715 PubMed Links

Gene Details

Gene
LOC101929715
Gene Name
uncharacterized LOC101929715
There are no transcripts associated with this gene.

Gene
ODC1
Gene Name
ornithine decarboxylase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287188.1 1487 Missense Mutation TCG,TTG S180L NP_001274117.1
NM_001287189.1 1487 Missense Mutation TCG,TTG S309L NP_001274118.1
NM_001287190.1 1487 Missense Mutation TCG,TTG S309L NP_001274119.1
NM_002539.2 1487 Missense Mutation TCG,TTG S309L NP_002530.1
Gene
SNORA80B
Gene Name
small nucleolar RNA, H/ACA box 80B
There are no transcripts associated with this gene.

View Full Product Details