Product Details

SNP ID
rs144541374
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:196836813 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGGGTGTAATTTTATAATACAAA[C/G]ATATTACTGCTTTCTTATCCTGGCA
Phenotype
MIM: 611655
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PGAP1 PubMed Links

Gene Details

Gene
PGAP1
Gene Name
post-GPI attachment to proteins 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321099.1 7136 UTR 3 NP_001308028.1
NM_001321100.1 7136 UTR 3 NP_001308029.1
NM_024989.3 7136 UTR 3 NP_079265.2
XM_017004992.1 7136 UTR 3 XP_016860481.1
XM_017004993.1 7136 UTR 3 XP_016860482.1
XM_017004994.1 7136 UTR 3 XP_016860483.1

View Full Product Details