Product Details
- SNP ID
-
rs146905811
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:109546167 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCAATTATTTCTTCTTCCAAAAGT[A/C]TTCTCTTTTCTTCAAGCTTCATTCT
- Phenotype
-
MIM: 601181
MIM: 611737
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
RANBP2
PubMed Links
Gene Details
- Gene
- RANBP2
- Gene Name
- RAN binding protein 2
- Gene
- SEPT10
- Gene Name
- septin 10
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001321496.1 |
1611 |
Missense Mutation |
AGA,ATA |
R278I |
NP_001308425.1 |
NM_001321498.1 |
1611 |
Missense Mutation |
AGA,ATA |
R411I |
NP_001308427.1 |
NM_001321499.1 |
1611 |
Missense Mutation |
AGA,ATA |
R244I |
NP_001308428.1 |
NM_001321500.1 |
1611 |
Missense Mutation |
AGA,ATA |
R388I |
NP_001308429.1 |
NM_001321501.1 |
1611 |
Missense Mutation |
AGA,ATA |
R369I |
NP_001308430.1 |
NM_001321502.1 |
1611 |
UTR 3 |
|
|
NP_001308431.1 |
NM_001321503.1 |
1611 |
UTR 3 |
|
|
NP_001308432.1 |
NM_001321504.1 |
1611 |
Missense Mutation |
AGA,ATA |
R218I |
NP_001308433.1 |
NM_001321505.1 |
1611 |
Missense Mutation |
AGA,ATA |
R218I |
NP_001308434.1 |
NM_001321506.1 |
1611 |
Missense Mutation |
AGA,ATA |
R221I |
NP_001308435.1 |
NM_001321507.1 |
1611 |
Missense Mutation |
AGA,ATA |
R278I |
NP_001308436.1 |
NM_001321508.1 |
1611 |
UTR 3 |
|
|
NP_001308437.1 |
NM_001321509.1 |
1611 |
Missense Mutation |
AGA,ATA |
R369I |
NP_001308438.1 |
NM_001321510.1 |
1611 |
Missense Mutation |
AGA,ATA |
R244I |
NP_001308439.1 |
NM_001321511.1 |
1611 |
Missense Mutation |
AGA,ATA |
R221I |
NP_001308440.1 |
NM_001321512.1 |
1611 |
Missense Mutation |
AGA,ATA |
R411I |
NP_001308441.1 |
NM_001321513.1 |
1611 |
Missense Mutation |
AGA,ATA |
R402I |
NP_001308442.1 |
NM_001321514.1 |
1611 |
Missense Mutation |
AGA,ATA |
R396I |
NP_001308443.1 |
NM_001321515.1 |
1611 |
Missense Mutation |
AGA,ATA |
R379I |
NP_001308444.1 |
NM_144710.4 |
1611 |
Missense Mutation |
AGA,ATA |
R411I |
NP_653311.1 |
NM_178584.3 |
1611 |
Missense Mutation |
AGA,ATA |
R388I |
NP_848699.1 |
XM_006712317.1 |
1611 |
Missense Mutation |
AGA,ATA |
R411I |
XP_006712380.1 |
XM_011510698.2 |
1611 |
Missense Mutation |
AGA,ATA |
R396I |
XP_011509000.1 |
XM_011510699.2 |
1611 |
Missense Mutation |
AGA,ATA |
R388I |
XP_011509001.1 |
XM_011510700.2 |
1611 |
Missense Mutation |
AGA,ATA |
R369I |
XP_011509002.1 |
XM_011510701.2 |
1611 |
Missense Mutation |
AGA,ATA |
R278I |
XP_011509003.1 |
XM_011510702.2 |
1611 |
Missense Mutation |
AGA,ATA |
R244I |
XP_011509004.1 |
XM_011510703.2 |
1611 |
Intron |
|
|
XP_011509005.1 |
XM_011510704.2 |
1611 |
UTR 3 |
|
|
XP_011509006.1 |
View Full Product Details