Product Details

SNP ID
rs146949759
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:10912534 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGTGGGGGGCGTGCGGCAGGTGCT[G/T]TACGGGGACCTCCTCAGTCAGTACC
Phenotype
MIM: 603787
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
KCNF1 PubMed Links

Gene Details

Gene
KCNF1
Gene Name
potassium voltage-gated channel modifier subfamily F member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002236.4 598 Silent Mutation CTG,CTT L36L NP_002227.2

View Full Product Details