Product Details
- SNP ID
-
rs148038387
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:241106634 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGCCATTAGGAGCCTCGCCTGGAA[C/T]GGGGCCCCCACAAAGCTCCTGAGCC
- Phenotype
-
MIM: 615393
MIM: 607505
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MTERF4
PubMed Links
Gene Details
- Gene
- MTERF4
- Gene Name
- mitochondrial transcription termination factor 4
There are no transcripts associated with this gene.
- Gene
- PASK
- Gene Name
- PAS domain containing serine/threonine kinase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001252119.1 |
4199 |
Missense Mutation |
ATT,GTT |
I1309V |
NP_001239048.1 |
NM_001252120.1 |
4199 |
Missense Mutation |
ATT,GTT |
I1302V |
NP_001239049.1 |
NM_001252122.1 |
4199 |
Missense Mutation |
ATT,GTT |
I1267V |
NP_001239051.1 |
NM_001252124.1 |
4199 |
Intron |
|
|
NP_001239053.1 |
NM_015148.3 |
4199 |
Missense Mutation |
ATT,GTT |
I1302V |
NP_055963.2 |
XM_005246991.1 |
4199 |
Missense Mutation |
ATT,GTT |
I1086V |
XP_005247048.1 |
XM_011510826.1 |
4199 |
Missense Mutation |
ATT,GTT |
I1334V |
XP_011509128.1 |
XM_011510828.1 |
4199 |
Missense Mutation |
ATT,GTT |
I1309V |
XP_011509130.1 |
XM_011510829.1 |
4199 |
Missense Mutation |
ATT,GTT |
I1309V |
XP_011509131.1 |
XM_011510830.1 |
4199 |
Missense Mutation |
ATT,GTT |
I1309V |
XP_011509132.1 |
XM_011510831.2 |
4199 |
Missense Mutation |
ATT,GTT |
I1123V |
XP_011509133.1 |
XM_011510832.2 |
4199 |
Missense Mutation |
ATT,GTT |
I1116V |
XP_011509134.1 |
XM_011510833.1 |
4199 |
Missense Mutation |
ATT,GTT |
I1093V |
XP_011509135.1 |
XM_011510834.1 |
4199 |
Missense Mutation |
ATT,GTT |
I923V |
XP_011509136.1 |
XM_011510835.1 |
4199 |
Missense Mutation |
ATT,GTT |
I852V |
XP_011509137.1 |
XM_017003634.1 |
4199 |
Missense Mutation |
ATT,GTT |
I1393V |
XP_016859123.1 |
XM_017003635.1 |
4199 |
Missense Mutation |
ATT,GTT |
I1302V |
XP_016859124.1 |
XM_017003636.1 |
4199 |
Missense Mutation |
ATT,GTT |
I1302V |
XP_016859125.1 |
View Full Product Details