Product Details

SNP ID
rs148059470
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:14634857 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCGACCTCGTGGACGACAAGGAGT[A/G]GCCGCCTAGGGGCTGCCGGCCCCTC
Phenotype
MIM: 611234
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM84A PubMed Links

Gene Details

Gene
FAM84A
Gene Name
family with sequence similarity 84 member A
There are no transcripts associated with this gene.

Gene
LOC653602
Gene Name
uncharacterized LOC653602
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291410.1 Intron NP_001278339.1

View Full Product Details