Product Details

SNP ID
rs148339452
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:172431750 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCCAAAACGGGCAGCGAGACTTA[C/T]AGGAAGTCTTAGGACTCTGCAAGAA
Phenotype
MIM: 147556
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ITGA6 PubMed Links

Gene Details

Gene
ITGA6
Gene Name
integrin subunit alpha 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000210.3 Intron NP_000201.2
NM_001079818.2 Intron NP_001073286.1
NM_001316306.1 Intron NP_001303235.1
XM_006712510.1 Intron XP_006712573.1
XM_006712511.1 Intron XP_006712574.1
XM_017004005.1 Intron XP_016859494.1
XM_017004006.1 Intron XP_016859495.1
XM_017004007.1 Intron XP_016859496.1
XM_017004008.1 Intron XP_016859497.1

View Full Product Details