Product Details
- SNP ID
-
rs149631076
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:110749956 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCAGGTGTTTGTCAAATGTCTCTCA[A/G]TTTGGGTTTGTCTGTGTTTTCCGTA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ACOXL
PubMed Links
Gene Details
- Gene
- ACOXL
- Gene Name
- acyl-CoA oxidase-like
View Full Product Details