Product Details

SNP ID
rs149800367
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:176189961 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGATAGCCAACTGCTTGCACCTGA[A/G]TGACACGCAAGTCAAAATCTGGTTC
Phenotype
MIM: 142987
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HAGLR PubMed Links

Gene Details

Gene
HAGLR
Gene Name
HOXD antisense growth-associated long non-coding RNA
There are no transcripts associated with this gene.

Gene
HOXD1
Gene Name
homeobox D1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024501.2 1070 Missense Mutation AAT,AGT N269S NP_078777.1
XM_005246507.3 1070 UTR 3 XP_005246564.1
Gene
MIR7704
Gene Name
microRNA 7704
There are no transcripts associated with this gene.

View Full Product Details