Product Details

SNP ID
rs150436368
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:55983072 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGACATCAAAAACTATGTAATAATT[A/G]TACTAAATATCATTTATTTCCAAAT
Phenotype
MIM: 610944 MIM: 615096
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC105374690 PubMed Links

Gene Details

Gene
LOC105374690
Gene Name
uncharacterized LOC105374690
There are no transcripts associated with this gene.

Gene
MIR216A
Gene Name
microRNA 216a
There are no transcripts associated with this gene.

Gene
MIR217
Gene Name
microRNA 217
There are no transcripts associated with this gene.

Gene
MIR217HG
Gene Name
MIR217 host gene
There are no transcripts associated with this gene.

View Full Product Details