Product Details

SNP ID
rs150844804
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:96267644 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACAGTGTGTAACCACTCTCGAGG[A/G]CCATGAAAATGAGGTCAAGTCAGTG
Phenotype
MIM: 604333 MIM: 601664 MIM: 613403
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CIAO1 PubMed Links

Gene Details

Gene
CIAO1
Gene Name
cytosolic iron-sulfur assembly component 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004804.2 513 Missense Mutation GAC,GGC D103G NP_004795.1
Gene
SNRNP200
Gene Name
small nuclear ribonucleoprotein U5 subunit 200
There are no transcripts associated with this gene.

Gene
TMEM127
Gene Name
transmembrane protein 127
There are no transcripts associated with this gene.

View Full Product Details