Product Details
- SNP ID
-
rs138162860
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:122328068 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAAGTATTAAGAACAGCTACCTCAC[C/T]CCAGAAAGGGGTTTTGCAGTTTTTA
- Phenotype
-
MIM: 184600
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CSTA
PubMed Links
Gene Details
- Gene
- CSTA
- Gene Name
- cystatin A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005213.3 |
|
Intron |
|
|
NP_005204.1 |
View Full Product Details