Product Details

SNP ID
rs138186262
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:146195186 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCACATCCAGGTCTAGTGGGAAGT[C/G]AATGTCAAAATGGTCAGCATCTGCC
Phenotype
MIM: 607612
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PLSCR4 PubMed Links

Gene Details

Gene
PLSCR4
Gene Name
phospholipid scramblase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128304.1 1124 Missense Mutation CAC,GAC H295D NP_001121776.1
NM_001128305.1 1124 Missense Mutation CAC,GAC H295D NP_001121777.1
NM_001128306.1 1124 Missense Mutation CAC,GAC H205D NP_001121778.1
NM_001177304.1 1124 Missense Mutation CAC,GAC H190D NP_001170775.1
NM_020353.2 1124 Missense Mutation CAC,GAC H295D NP_065086.2
XM_005247654.2 1124 Missense Mutation CAC,GAC H295D XP_005247711.1
XM_005247655.2 1124 Missense Mutation CAC,GAC H295D XP_005247712.1
XM_011513031.2 1124 Missense Mutation CAC,GAC H295D XP_011511333.1
XM_017006921.1 1124 Missense Mutation CAC,GAC H205D XP_016862410.1

View Full Product Details