Product Details

SNP ID
rs139319109
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:101685871 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGAGGACAGTCCAGTTTATCTGCC[A/G]AGGATTCCTCTTGGAAAGGAAAGCC
Phenotype
MIM: 604180
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC105374013 PubMed Links

Gene Details

Gene
LOC105374013
Gene Name
endogenous retrovirus group K member 5 Gag polyprotein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011513341.2 Intron XP_011511643.1
Gene
RPL24
Gene Name
ribosomal protein L24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000986.3 Intron NP_000977.1
Gene
ZBTB11
Gene Name
zinc finger and BTB domain containing 11
There are no transcripts associated with this gene.

Gene
ZBTB11-AS1
Gene Name
ZBTB11 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details