Product Details

SNP ID
rs139601257
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:32528405 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGAACTTCATTATATCTCAAAAAA[C/G]CTGTTATTTTAAACAAGTGACTTGC
Phenotype
MIM: 615890
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DYNC1LI1 PubMed Links

Gene Details

Gene
DYNC1LI1
Gene Name
dynein cytoplasmic 1 light intermediate chain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016141.3 Intron NP_057225.2

View Full Product Details