Product Details

SNP ID
rs139680303
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:10249369 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCAGCAGCCCCAAGCGCCTGAAA[A/G]CCCAGAAGGAGGACGATGTGGCTTG
Phenotype
MIM: 603304
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
IRAK2 PubMed Links

Gene Details

Gene
IRAK2
Gene Name
interleukin 1 receptor associated kinase 2
There are no transcripts associated with this gene.

Gene
TATDN2
Gene Name
TatD DNase domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014760.3 750 Missense Mutation ACC,GCC T57A NP_055575.3

View Full Product Details