Product Details

SNP ID
rs141128323
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:195748909 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCACAGCCCTATGCACCTGCAGC[A/G]GGGCCCACTGGGCAGGTGCTGGCAC
Phenotype
MIM: 158372
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MUC4 PubMed Links

Gene Details

Gene
MUC4
Gene Name
mucin 4, cell surface associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322468.1 22201 Missense Mutation NP_001309397.1
NM_004532.5 22201 Missense Mutation NP_004523.3
NM_018406.6 22201 Intron NP_060876.5
NM_138297.4 22201 Missense Mutation NP_612154.2

View Full Product Details