Product Details

SNP ID
rs141668972
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:28322855 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATCTGAGATATTGAGTTCAAGATA[C/T]ATGAAACTGTCTATCAAAAAGGATA
Phenotype
MIM: 609916 MIM: 615166
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AZI2 PubMed Links

Gene Details

Gene
AZI2
Gene Name
5-azacytidine induced 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134432.1 2438 Intron NP_001127904.1
NM_001134433.1 2438 Intron NP_001127905.1
NM_001271650.1 2438 UTR 3 NP_001258579.1
NM_022461.4 2438 UTR 3 NP_071906.1
XM_005265392.2 2438 UTR 3 XP_005265449.1
XM_011534009.2 2438 UTR 3 XP_011532311.1
XM_011534010.2 2438 Intron XP_011532312.1
XM_011534011.2 2438 Intron XP_011532313.1
Gene
CMC1
Gene Name
C-X9-C motif containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182523.1 2438 Intron NP_872329.1
XM_011533402.2 2438 UTR 3 XP_011531704.1
XM_017005760.1 2438 Intron XP_016861249.1

View Full Product Details