Product Details

SNP ID
rs143152801
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:58517232 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATCAGGCGGAGCAGGTCCAGGTA[G/T]GCTGTTCTTGCCATGTCCACTTGGG
Phenotype
MIM: 601641
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ACOX2 PubMed Links

Gene Details

Gene
ACOX2
Gene Name
acyl-CoA oxidase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003500.3 2001 Silent Mutation NP_003491.1
XM_005265505.1 2001 Silent Mutation XP_005265562.1
XM_006713340.3 2001 Silent Mutation XP_006713403.1
XM_017007302.1 2001 Silent Mutation XP_016862791.1

View Full Product Details