Product Details
- SNP ID
-
rs143361027
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:126008759 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGATCAGGCCTGCCAGCAGGTAGAG[C/G]ACCACAAAGGTCTGGCTGTTTATGA
- Phenotype
-
MIM: 610803
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
SLC41A3
PubMed Links
Gene Details
- Gene
- SLC41A3
- Gene Name
- solute carrier family 41 member 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001008485.1 |
1376 |
Silent Mutation |
GTC,GTG |
V409V |
NP_001008485.1 |
NM_001008486.1 |
1376 |
Silent Mutation |
GTC,GTG |
V373V |
NP_001008486.1 |
NM_001008487.1 |
1376 |
Silent Mutation |
GTC,GTG |
V383V |
NP_001008487.1 |
NM_001164475.1 |
1376 |
Silent Mutation |
GTC,GTG |
V292V |
NP_001157947.1 |
NM_017836.3 |
1376 |
Silent Mutation |
GTC,GTG |
V409V |
NP_060306.3 |
XM_005247559.1 |
1376 |
Silent Mutation |
GTC,GTG |
V424V |
XP_005247616.1 |
XM_005247562.1 |
1376 |
Silent Mutation |
GTC,GTG |
V409V |
XP_005247619.1 |
XM_005247563.2 |
1376 |
Silent Mutation |
GTC,GTG |
V409V |
XP_005247620.1 |
XM_005247564.1 |
1376 |
Silent Mutation |
GTC,GTG |
V424V |
XP_005247621.1 |
XM_005247565.2 |
1376 |
Silent Mutation |
GTC,GTG |
V383V |
XP_005247622.1 |
XM_006713681.2 |
1376 |
Silent Mutation |
GTC,GTG |
V409V |
XP_006713744.1 |
XM_011512945.1 |
1376 |
Silent Mutation |
GTC,GTG |
V398V |
XP_011511247.1 |
XM_017006705.1 |
1376 |
Silent Mutation |
GTC,GTG |
V424V |
XP_016862194.1 |
XM_017006706.1 |
1376 |
Silent Mutation |
GTC,GTG |
V409V |
XP_016862195.1 |
XM_017006707.1 |
1376 |
Silent Mutation |
GTC,GTG |
V409V |
XP_016862196.1 |
XM_017006708.1 |
1376 |
Silent Mutation |
GTC,GTG |
V373V |
XP_016862197.1 |
XM_017006709.1 |
1376 |
Silent Mutation |
GTC,GTG |
V373V |
XP_016862198.1 |
View Full Product Details