Product Details

SNP ID
rs146210920
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:52814262 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGTCAGTGTCACGCAGAAGGAGC[C/T]GGAGCCCCTCCCCACGGCCATCCCA
Phenotype
MIM: 146650 MIM: 600564
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ITIH3 PubMed Links

Gene Details

Gene
ITIH3
Gene Name
inter-alpha-trypsin inhibitor heavy chain 3
There are no transcripts associated with this gene.

Gene
ITIH4
Gene Name
inter-alpha-trypsin inhibitor heavy chain family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166449.1 2542 Missense Mutation CAG,CGG Q828R NP_001159921.1
NM_002218.4 2542 Missense Mutation CAG,CGG Q858R NP_002209.2
Gene
ITIH4-AS1
Gene Name
ITIH4 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details