Product Details

SNP ID
rs147545399
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:14126546 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTAGCTGTGTGGCTTTCATCTCT[C/T]CAGGTTTATGTAGCCTCATGGATCA
Phenotype
MIM: 611077 MIM: 612048
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHCHD4 PubMed Links

Gene Details

Gene
CHCHD4
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 4
There are no transcripts associated with this gene.

Gene
TMEM43
Gene Name
transmembrane protein 43
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024334.2 Intron NP_077310.1
XM_017007176.1 Intron XP_016862665.1

View Full Product Details