Product Details

SNP ID
rs149679373
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:138495021 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAATGATTTGATTTGTTTTCAGTA[G/T]GAAAGTACTTTTAATTTCTTTACTG
Phenotype
MIM: 614310
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CEP70 PubMed Links

Gene Details

Gene
CEP70
Gene Name
centrosomal protein 70
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288964.1 2418 Silent Mutation NP_001275893.1
NM_001288965.1 2418 Silent Mutation NP_001275894.1
NM_001288966.1 2418 Intron NP_001275895.1
NM_001288967.1 2418 Silent Mutation NP_001275896.1
NM_001320598.1 2418 Silent Mutation NP_001307527.1
NM_001320599.1 2418 Intron NP_001307528.1
NM_001320600.1 2418 Intron NP_001307529.1
NM_024491.3 2418 Silent Mutation NP_077817.2
XM_005247805.3 2418 Intron XP_005247862.1
XM_011513209.2 2418 Intron XP_011511511.1
XM_017007270.1 2418 Intron XP_016862759.1
XM_017007271.1 2418 Intron XP_016862760.1
XM_017007272.1 2418 Intron XP_016862761.1
XM_017007273.1 2418 Intron XP_016862762.1
XM_017007274.1 2418 Intron XP_016862763.1
XM_017007275.1 2418 Silent Mutation XP_016862764.1
XM_017007276.1 2418 Intron XP_016862765.1
XM_017007277.1 2418 Intron XP_016862766.1
XM_017007278.1 2418 Intron XP_016862767.1
XM_017007279.1 2418 Intron XP_016862768.1
XM_017007280.1 2418 Intron XP_016862769.1
XM_017007281.1 2418 Intron XP_016862770.1
XM_017007282.1 2418 Silent Mutation XP_016862771.1
XM_017007283.1 2418 Silent Mutation XP_016862772.1
XM_017007284.1 2418 Silent Mutation XP_016862773.1

View Full Product Details