Product Details

SNP ID
rs149822536
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:146194000 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCTTCAGTATGAGGACATGTTGA[C/T]GGAGAGGAAAGGTAGGAAAGGGTTA
Phenotype
MIM: 607612
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLSCR4 PubMed Links

Gene Details

Gene
PLSCR4
Gene Name
phospholipid scramblase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128304.1 1642 UTR 3 NP_001121776.1
NM_001128305.1 1642 UTR 3 NP_001121777.1
NM_001128306.1 1642 UTR 3 NP_001121778.1
NM_001177304.1 1642 UTR 3 NP_001170775.1
NM_020353.2 1642 UTR 3 NP_065086.2
XM_005247654.2 1642 UTR 3 XP_005247711.1
XM_005247655.2 1642 UTR 3 XP_005247712.1
XM_011513031.2 1642 UTR 3 XP_011511333.1
XM_017006921.1 1642 UTR 3 XP_016862410.1

View Full Product Details