Product Details

SNP ID
rs150294581
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:49271382 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTTGGCAATGACCACACATGTGG[C/T]TCAATTCGTTTTCAAATTCTATTTT
Phenotype
MIM: 603486
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C3orf62 PubMed Links

Gene Details

Gene
C3orf62
Gene Name
chromosome 3 open reading frame 62
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198562.2 805 Missense Mutation AAC,AGC N201S NP_940964.1
Gene
MIR4271
Gene Name
microRNA 4271
There are no transcripts associated with this gene.

Gene
USP4
Gene Name
ubiquitin specific peptidase 4
There are no transcripts associated with this gene.

View Full Product Details