Product Details

SNP ID
rs150659806
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:27717080 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTTAAAATAACTCTTTAGGGAGTT[A/G]TGTAAAAAGCATAATACCCTCCCAT
Phenotype
MIM: 604615
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EOMES PubMed Links

Gene Details

Gene
EOMES
Gene Name
eomesodermin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278182.1 2408 Missense Mutation ACA,ATA T703I NP_001265111.1
NM_001278183.1 2408 Missense Mutation ACA,ATA T408I NP_001265112.1
NM_005442.3 2408 Missense Mutation ACA,ATA T684I NP_005433.2
XM_005265510.3 2408 UTR 3 XP_005265567.1

View Full Product Details