Product Details

SNP ID
rs151056679
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:180984710 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGGCTGCTATATAAGGAGATCCT[A/C]CTATAGGAAGAAAGAAAAAAGAACA
Phenotype
MIM: 608977 MIM: 600819
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DNAJC19 PubMed Links

Gene Details

Gene
DNAJC19
Gene Name
DnaJ heat shock protein family (Hsp40) member C19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190233.1 453 Missense Mutation GGA,GTA G69V NP_001177162.1
NM_145261.3 453 Missense Mutation GGA,GTA G94V NP_660304.1
Gene
FXR1
Gene Name
FMR1 autosomal homolog 1
There are no transcripts associated with this gene.

View Full Product Details