Product Details

SNP ID
rs138450883
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:133150872 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACCATCCGAGTGCTGGACTCCAA[C/T]GACAATGTGCCCGCTTTCGACCAAC
Phenotype
MIM: 608286
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101927359 PubMed Links

Gene Details

Gene
LOC101927359
Gene Name
uncharacterized LOC101927359
There are no transcripts associated with this gene.

Gene
PCDH10
Gene Name
protocadherin 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020815.2 1583 Silent Mutation AAC,AAT N244N NP_065866.1
NM_032961.2 1583 Silent Mutation AAC,AAT N244N NP_116586.1
XM_011532150.1 1583 Silent Mutation AAC,AAT N244N XP_011530452.1

View Full Product Details