Product Details

SNP ID
rs138708352
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:154234587 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAGTTCATGGCATGTACCACTGAT[A/G]TGTGTTCCTAATAATTCTCCTTCTC
Phenotype
MIM: 612486
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DCHS2 PubMed Links

Gene Details

Gene
DCHS2
Gene Name
dachsous cadherin-related 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142552.1 5994 Intron NP_001136024.1
NM_017639.3 5994 Silent Mutation CAC,CAT H2900H NP_060109.2
XM_011532045.1 5994 Silent Mutation CAC,CAT H3355H XP_011530347.1
XM_011532046.2 5994 Silent Mutation CAC,CAT H2578H XP_011530348.1
XM_011532049.2 5994 Silent Mutation CAC,CAT H1690H XP_011530351.1
XM_017008320.1 5994 Intron XP_016863809.1

View Full Product Details