Product Details
- SNP ID
-
rs139967072
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:163471861 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTAGAAATAAGCCTAGTTTACTTC[A/G]TTAAAGTAAGTGTTACGGCTGCTAT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TKTL2
PubMed Links
Gene Details
- Gene
- TKTL2
- Gene Name
- transketolase like 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_032136.4 |
2035 |
Missense Mutation |
ACG,ATG |
T625M |
NP_115512.3 |
View Full Product Details