Product Details

SNP ID
rs145063393
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:70689137 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGAGGCCGGCAGAGTCAACAGGAG[G/T]CAGAGGAGGAGGAAAGAGAGGAGGA
Phenotype
MIM: 611614
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
UTP3 PubMed Links

Gene Details

Gene
UTP3
Gene Name
UTP3, small subunit processome component homolog (S. cerevisiae)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020368.2 659 Missense Mutation GCA,TCA A154S NP_065101.1

View Full Product Details