Product Details

SNP ID
rs146492023
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:155345344 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTGAATGACTTGTTGGGTCTATG[C/T]AAACACCTCATCCTACCTCAGAAGC
Phenotype
MIM: 610070
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC102724776 PubMed Links

Gene Details

Gene
LOC102724776
Gene Name
uncharacterized LOC102724776
There are no transcripts associated with this gene.

Gene
MAP9
Gene Name
microtubule associated protein 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039580.1 4664 UTR 3 NP_001034669.1
XM_006714306.3 4664 UTR 3 XP_006714369.1
XM_011532253.1 4664 UTR 3 XP_011530555.1
XM_011532254.2 4664 UTR 3 XP_011530556.1
XM_011532255.2 4664 UTR 3 XP_011530557.1
XM_017008616.1 4664 UTR 3 XP_016864105.1
XM_017008617.1 4664 UTR 3 XP_016864106.1
XM_017008618.1 4664 Intron XP_016864107.1

View Full Product Details