Product Details

SNP ID
rs146710132
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:177434395 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAAGTCATACCTTGGCAGGAAGC[A/G]CATCAATATATCACCATTCCCAGTG
Phenotype
MIM: 613228
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AGA PubMed Links

Gene Details

Gene
AGA
Gene Name
aspartylglucosaminidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000027.3 891 Missense Mutation CGC,TGC R265C NP_000018.2
NM_001171988.1 891 Missense Mutation CGC,TGC R255C NP_001165459.1
Gene
LOC285500
Gene Name
uncharacterized LOC285500
There are no transcripts associated with this gene.

View Full Product Details